Learn More
Medchemexpress LLC Biotin-PEG2-C2-iodoacetamide | 292843-75-5 | 98.2% | C18H31IN4O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140941100MG
Encompass_Preferred
Biotin-PEG2-C2-iodoacetamide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-53174