Learn More
Medchemexpress LLC Biotin-PEG2-C2-iodoacetamide | 292843-75-5 | 98.2% | C18H31IN4O5S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140941
Encompass_Preferred
Biotin-PEG2-C2-iodoacetamide is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are compounds featuring two distinct ligands linked by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets a specific protein. These molecules leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Utilizes the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-57377