Learn More
Medchemexpress LLC Biotin-PEG2-C6-azide 5mg | 1011268-29-3 | 513.65 g·mol-1 | C22H39N7O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1170425MG
Encompass_Preferred
Biotin-PEG2-C6-azide is a biotinylated azide reagent used as a click-chemistry linker for bioconjugation, including PROTAC assembly, probe labeling, and affinity capture. The reagent features a biotin tag connected via a PEG2 spacer and a C6 alkyl spacer terminating in an azide for copper-catalyzed or strain-promoted azide-alkyne cycloaddition. Molecular formula C22H39N7O5S; molecular weight ≈ 513.65 g·mol-1.
  • Provides a biotin handle for streptavidin/avidin affinity capture.
  • Contains a PEG2 spacer to improve solubility and flexibility.
  • Features a C6 linker terminating in an azide for efficient click chemistry conjugation.
  • Suitable for PROTAC linker synthesis, probe labeling, and other bioconjugation workflows.
  • Typically stored frozen to preserve reagent integrity (store at -20°C).
Catalog No. 50-002-29910