Learn More
Medchemexpress LLC Biotin-PEG2-Mal | 305372-39-8 | 99.6% | 525.62 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW010764
Encompass_Preferred
Biotin-PEG2-Mal is a PEG-based PROTAC linker that can be used in the synthesis of PROTAC. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in protac synthesis
  • Facilitates degradation of target proteins
  • Exploits the ubiquitin-proteasome system
Catalog No. 50-004-51523