Learn More
Aat Bioquest Biotin PEG2 maleimide CAS 305372-39-8 (5 mg)
SDP
Supplier:  Aat Bioquest 3015
Encompass_Preferred
This thiol-reactive biotin derivative contains a long arm 20 angstrom to increase its avidin-binding affinity. This catalog is for 5 mg.
Catalog No. 50-224-9383