Learn More
Medchemexpress LLC Biotin-PEG3-alcohol | 1263044-40-1 | >97.0% | C16H29N3O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13592410MG
Encompass_Preferred
Biotin-PEG3-OH is a biotinylated, hydroxy-terminated triethylene glycol (PEG3) linker used to introduce a biotin affinity tag into small molecules, peptides, and PROTAC constructs for chemical biology research. It is supplied as a waxy solid and is commonly used in affinity labeling, pull-down assays, and conjugation chemistry; follow supplier guidance for cold, dry storage.
  • Provides biotin tag for streptavidin-based capture
  • Peg3 spacer improves solubility and reduces steric hindrance
  • Terminal hydroxyl enables further chemical modification
  • Suitable for PROTAC linker synthesis and conjugation
  • Typical purity >97.0% and stable when stored at -20 °C
Catalog No. 50-002-59191