Learn More
Cayman Chemical -BIotIn-PEG3-amIn 100mg
SDP
Supplier:  Cayman Chemical 30534100 MG
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of biotinylated siderophores for whole-cell bacterial pull-down assays, as well as in the synthesis of protein-polysaccharide conjugate vaccines for S. pneumoniae
Catalog No. 50-299-0649