Learn More
Cayman Chemical -BIotIn-PEG3-amIn 50mg
SDP
Supplier:  Cayman Chemical 3053450 MG
Encompass_Preferred
A fluorescent probe abs/em 346/376 nm respectively fluorescence can be quenched by ferric ions and the nitroaromatic compounds and explosive components 24-DNT and TNT in aqueous solutions fluorescence quenching has been used in study of reverse micelles including intermicellar migration of reactants and in the estimation of the number of micelles per cluster has also been used as a surfactant and dopant in the synthesis of polypyrrole micro- and nanowires
Catalog No. 50-298-9600