Learn More
Cayman Chemical BIotIn-PEG3-azIde 5mg
SDP
Supplier:  Cayman Chemical 234195 MG
Encompass_Preferred
A PEGylated version of biotin-azide that is more hydrophilic; can be used to label terminal alkynes via copper-catalyzed click reactions or copper-free click reactions with cyclooctynes; used to covalently tag proteins in combination with other tags
Catalog No. 50-296-3926