Learn More
Medchemexpress LLC Biotin-PEG3-azide | 875770-34-6 | ≥97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130143100MG
Encompass_Preferred
Biotin-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Can be conjugated to antiviral inhibitors
  • Exhibits broad-spectrum inhibition against various pathogenic viruses
  • Effective against hepatitis C virus, dengue virus, Zika virus, chikungunya virus, and enterovirus
Catalog No. 50-003-39040