Learn More
Medchemexpress LLC Biotin-PEG3-azide | 875770-34-6 | ≥97.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13014350MG
Encompass_Preferred
Biotin-PEG3-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which facilitates specific chemical reactions.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-40007