Learn More
Medchemexpress LLC Biotin-PEG3-bromide | 1041766-91-9 | 95.0% | 482.43 g/mol | C18H32BrN3O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101974100MG
Encompass_Preferred
Biotin-PEG3-bromide is a biotinylated PEG3 linker terminating in a reactive bromide. It provides a short, hydrophilic spacer for biotin conjugation and affinity capture, while the terminal bromide can be displaced by nucleophiles or used in cross-coupling reactions to install diverse handles. The reagent is intended for pull-down assays, streptavidin-mediated capture, and small-molecule bioconjugation workflows.
Catalog No. 50-002-47992