Learn More
Medchemexpress LLC Biotin-PEG3-NHS ester | 1253286-56-4 | 98.4% | 50 MG
Supplier: Medchemexpress LLC HY13029150MG
Biotin-PEG3-NHS ester is a PEG-based PROTAC linker, specifically designed for use in the synthesis of PROTACs. Proteolysis Targeting Chimeras (PROTACs) function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound exhibits a high purity of 98.35%, has a molecular weight of 544.62, and the chemical formula C23H36N4O9S. It is presented as a solid, ranging in color from white to light yellow.
- Functions as a PEG-based linker for the synthesis of PROTACs.
- Enables PROTACs to exploit the intracellular ubiquitin-proteasome system for targeted protein degradation.
- Offers a high purity of 98.35%.
- Molecular weight: 544.62; formula: C23H36N4O9S; CAS No.: 1253286-56-4.
- Available as a solid, with a color ranging from white to light yellow.