Learn More
Medchemexpress LLC Biotin-PEG4-acid | 721431-18-1 | 97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126959100MG
Encompass_Preferred
Biotin-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are compounds with two ligands connected by a linker; one binds to an E3 ubiquitin ligase, and the other targets a specific protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Can be utilized in the synthesis of protacs
  • Has a purity of 97.0%
  • Appears as a white to off-white solid
  • Intended for research use only
Catalog No. 50-003-40882