Learn More
Medchemexpress LLC Biotin-PEG4-alcohol | 1217609-84-1 | 98.0% | C18H33N3O6S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13029010MG
Encompass_Preferred
Biotin-PEG4-OH is a biotinylated polyethylene glycol linker with a terminal hydroxyl group, designed for use as a flexible spacer and affinity tag in chemical biology and PROTAC synthesis. The PEG4 spacer increases solubility and reduces steric hindrance while enabling conjugation to biomolecules and small-molecule ligands.
Catalog No. 50-002-60766