Learn More
Medchemexpress LLC Biotin-PEG4-alkyne | 1458576-00-5 | 99.7% | C21H35N3O6S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14092250MG
Encompass_Preferred
Biotin-PEG4-alkyne is a biotin-labeled, PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess Azide groups. This product is intended for research use only and is not sold to patients.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Supports selective degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-55774