Learn More
Medchemexpress LLC Biotin-PEG4-alkyne | 1458576-00-5 | MFCD22380755 | 99.7% | 457.58 g/mol | C21H35N3O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1409225MG
Encompass_Preferred
Biotin-PEG4-alkyne is a biotin-labeled polyethylene glycol (PEG4) linker with a terminal alkyne designed for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It enables efficient biotinylation and affinity tagging of azide-containing molecules for pull-downs, imaging, or PROTAC assembly, offering good solubility and high chemical purity for biochemical applications.
  • Terminal alkyne for CuAAC click chemistry.
  • PEG4 spacer to improve solubility and reduce steric hindrance.
  • Biotin handle for strong streptavidin/avidin affinity capture.
  • High purity suitable for analytical and preparative workflows.
  • Suitable for bioconjugation, labeling, and PROTAC linker assembly.
Catalog No. 50-002-23136