Learn More
Medchemexpress LLC Biotin-PEG4-amide-alkyne | 1006592-45-5 | MFCD16621136 | 95.0% | C24H40N4O7S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14092310MG
Encompass_Preferred
Biotin-PEG4-amide-alkyne is a polyethylene glycol (PEG)-based linker reagent that combines a biotin affinity tag with a terminal alkyne group for click chemistry applications. Supplied as an off-white to light yellow solid (CAS 1006592-45-5), it is designed for bioconjugation, affinity capture, and incorporation into PROTAC and probe chemistries.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Includes a biotin moiety for strong affinity capture and detection.
  • Features a PEG4 spacer to enhance aqueous solubility and reduce steric hindrance.
  • Supplied as a stable solid suitable for storage and handling in research labs.
  • Applicable to bioconjugation, probe synthesis, and PROTAC linker construction.
Catalog No. 50-002-59694