Learn More
Medchemexpress LLC Biotin-PEG4-amine | 663171-32-2 | 98.0% | C20H38N4O6S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140895100MG
Encompass_Preferred
Biotin-PEG4-amine is a biotin-labeled, PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled, PEG-based linker
  • Used for the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55404