Learn More
Medchemexpress LLC Biotin-PEG4-amine | 663171-32-2 | MFCD26142992 | 98.0% | C20H38N4O6S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14089550MG
Encompass_Preferred
Biotin-PEG4-amine is a biotin-labeled, PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled for specific applications
  • PEG-based for improved solubility and reduced immunogenicity
  • Functions as a PROTAC linker
  • Facilitates targeted protein degradation
Catalog No. 50-003-55795