Learn More
Medchemexpress LLC Biotin-PEG4-amino-t-Bu-DADPS-C3-alykne | 2241685-22-1 | 95.0% | C42H62N4O9SSi | 10MG
SDP
Supplier:  Medchemexpress LLC HY14092810MG
Encompass_Preferred
Biotin-PEG4-amino-t-Bu-DADPS-C3-alykne is a PEG-based bioconjugation linker combining a biotin tag, a PEG4 spacer, a primary amino handle, a DADPS cleavable protecting group, and a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is designed for use in PROTAC assembly, affinity capture, and click-chemistry-based labeling workflows.
  • Contains a terminal alkyne for CuAAC click chemistry.
  • Includes a biotin moiety for affinity capture or detection.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Primary amino handle enables amide coupling to carboxyl-containing partners.
  • DADPS group provides acid-cleavable protection for controlled release.
  • Stable as a powder when stored cold and protected from light.
Catalog No. 50-002-60365