Learn More
TARGETMOL CHEMICALS INC BIOTIN-PEG4-AZIDE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T17583100MG
Encompass_Preferred
Also available in 25 mg 50 mg and bulk. Please contact Fisher for quotes. Biotin-PEG4-azide is a biotin-labeled PEG-based PROTAC linker that can be used in PROTAC synthesis. purity: 99%
Catalog No. 50-258-1100