Learn More
Medchemexpress LLC Biotin-PEG4-MeTz | 1962919-31-8 | C31H46N8O7S | 5 MG

Supplier: Medchemexpress LLC HY1517415MG
Biotin-PEG4-MeTz is a click chemistry reagent that contains a terminal methyltetrazine group, which reacts with trans-cyclooctene. It can be used to prepare biotinylated conjugates.
- Purity: 95.32%
- Molecular weight: 674.81
- Appearance: Solid
- Color: Purple to purplish red
- Storage: -20°C, sealed, away from moisture and light
- In solvent storage: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light)
- Solubility in DMSO: 250 mg/mL (370.47 mM), requires ultrasonication
- Designed for click chemistry applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.