Learn More
Medchemexpress LLC Biotin-PEG4-NHS ester | 459426-22-3 | 99.6% | C25H40N4O10S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140889
Encompass_Preferred
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs, which are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Biotin-labeled compound
  • PEG-based linker
  • Used for PROTAC synthesis
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52761