Learn More
Medchemexpress LLC Biotin-PEG4-NHS ester | 459426-22-3 | MFCD08458915 | 99.6% | C25H40N4O10S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140889500MG
Encompass_Preferred
Biotin-PEG4-NHS ester is a biotin-labeled, PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs, compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Designed for use in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • The other ligand binds to the target protein
Catalog No. 50-003-55164