Learn More
Medchemexpress LLC Biotin-PEG4-OH | 1217609-84-1 | 98.0% | 25 MG

Supplier: Medchemexpress LLC 25MGHY130290
Biotin-PEG4-OH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, with one ligand binding to an E3 ubiquitin ligase and the other to the target protein. These molecules leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. It has a molecular weight of 419.54 and an appearance of a white to off-white solid. This product is for research use only.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Leverages the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- Molecular weight: 419.54
- Appearance: white to off-white solid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.