Learn More
Medchemexpress LLC Biotin-PEG4-picolyl azide | 2222687-71-8 | MFCD29052186 | 98.7% | C27H42N8O7S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14091510MG
Encompass_Preferred
Biotin-PEG4-picolyl azide is a biotinylated PEG linker with a picolyl azide group designed for click chemistry and biochemical workflows. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), and the biotin moiety enables affinity capture, detection, and purification in assays.
  • Enables CuAAC reactions with alkyne-containing molecules.
  • Compatible with SPAAC reactions using DBCO or BCN partners.
  • Provides a biotin affinity tag for capture and detection workflows.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for biochemical applications.
Catalog No. 50-002-58375