Learn More
Medchemexpress LLC Biotin-PEG4-TFP ester | 2985664-57-9 | 97.8% | C27H37F4N3O8S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140903
Encompass_Preferred
Biotin-PEG4-TFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. This linker exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-52592