Learn More
Medchemexpress LLC Biotin-peg6-mal | 1808990-66-0 | 99.0% | C31H51N5O11S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14090810MG
Encompass_Preferred
Biotin-PEG6-Mal is a biotin-labeled polyethylene glycol (PEG6) maleimide linker that enables thiol-selective conjugation to introduce a biotin affinity tag for capture or detection using streptavidin/avidin systems. It is commonly used in PROTAC assembly and bioconjugation workflows where solubility and specific thiol reactivity are required.
  • Thiol-selective maleimide group for rapid, specific conjugation.
  • PEG6 spacer improves solubility and reduces steric hindrance.
  • Biotin affinity tag enables high-affinity streptavidin/avidin capture.
  • High purity supports reproducible experimental results.
  • Stable as a powdered reagent with defined cold-storage recommendations.
Catalog No. 50-002-60178