Learn More
Medchemexpress LLC Biotin-PEG6-NHS ester | 2055045-04-8 | MFCD22056308 | 98.0% | C29H48N4O12S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14089010MG
Encompass_Preferred
Biotin-PEG6-NHS ester is an amine-reactive, PEG-based linker containing a biotin tag and an N-hydroxysuccinimide (NHS) ester for labeling primary amines. It is commonly used to biotinylate proteins, peptides, and other amine-containing molecules and as a PEG-based linker in PROTAC synthesis to enable affinity capture or target engagement studies.
  • Amine-reactive NHS ester for efficient, selective bioconjugation.
  • PEG6 spacer provides a flexible, hydrophilic linker.
  • Biotin tag enables streptavidin-based affinity capture and detection.
  • High purity (98.0%) suitable for research applications.
  • Solid form; store sealed at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Compatible with protein and peptide labeling workflows.
Catalog No. 50-002-61537