Learn More
Medchemexpress LLC Biotin-PEG6-silane | 00-00-0 | 98.0% | 783.06 g/mol | C34H66N4O12SSi | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1384195MG
Encompass_Preferred
Biotin-PEG6-silane is a biotinylated PEG6 linker bearing a terminal silane group, intended for use in chemical biology applications such as PROTAC assembly, affinity capture, and surface functionalization. The PEG6 spacer positions the biotin tag while the silane enables surface coupling to silica and glass substrates, supporting conjugation and immobilization workflows.
  • Provides a biotin tag with a six-unit PEG spacer.
  • Terminal silane enables covalent attachment to silica and glass surfaces.
  • Suitable for PROTAC linker assembly and affinity capture workflows.
  • PEG spacer improves solubility and reduces steric hindrance.
  • High purity for reproducible experimental results.
Catalog No. 50-002-22857