Learn More
Medchemexpress LLC Biotin-PEG7-amine | 1334172-76-7 | 98.0% | C26H50N4O9S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140898
Encompass_Preferred
Biotin-PEG7-amine is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52857