Learn More
Medchemexpress LLC Biotin-PEG7-amine | 1334172-76-7 | 98.0% | C26H50N4O9S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14089850MG
Encompass_Preferred
Biotin-PEG7-amine is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs achieve this by connecting two different ligands via a linker, one binding to an E3 ubiquitin ligase and the other to a target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54563