Learn More
Medchemexpress LLC Biotin-PEG7-amine | 1334172-76-7 | MFCD21363308 | >95.0% | 594.76 g·mol⁻¹ | C26H50N4O9S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1408985MG
Encompass_Preferred
Biotin-PEG7-amine is a biotinylated polyethylene glycol (PEG7) reagent with a terminal primary amine. It serves as a flexible linker or labeling reagent for PROTAC synthesis and general bioconjugation, providing a biotin handle for affinity capture and detection while improving solubility and reducing steric hindrance.
  • Provides a biotin tag for affinity purification and detection.
  • Contains a PEG7 spacer to increase solubility and flexibility.
  • Features a terminal primary amine for amide coupling or NHS-ester reactions.
  • Appropriate for PROTAC linker construction and general bioconjugation workflows.
  • Offered in multiple pack sizes to suit research needs.
Catalog No. 50-002-23254