Learn More
AURORA BIOLABS LLC Biotin Protein Ligase (BirA), 100 μg
Supplier:  AURORA BIOLABS LLC 90101100
Encompass
Biotin-protein ligase activates biotin to form biotinyl 5' adenylate and transfers the biotin to biotin-accepting proteins. Biotin ligase also controls biotin synthesis in Escherichia coli. As a homodimer, it negatively controls the biotin synthetic operon.
Catalog No. NC3787975