Learn More
Seracare Life Sciences Inc BIOTIN-SA HRP KIT MS PRI AB
Supplier:  Seracare Life Sciences Inc 55200023
Encompass
HistoMark Biotin Streptavidin-HRP Systems provide rapid, precise localization of cell surface and intracellular antigens in frozen or paraffin-embedded tissue, cytospins and touch preparations. These systems also facilitate double or triple labeleing experiments, as detection can be performed simultaneously with primary antibodies of different animal species Streptavidin demonstrates less non-specific binding than avidin. Biotinylated anti-mouse antibody is human serum adsorbed to minimize cross-reactivity.
Catalog No. 50-675-65