Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-SLC | 1864003-57-5 | ≥97.0% | 100 MG
SDP

Supplier:  Medchemexpress LLC HY130887100MG

Encompass_Preferred

Biotin-SLC is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other for the target protein. They function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Alkyl chain-based PROTAC linker
  • Used for PROTAC synthesis
  • Exploits the intracellular ubiquitin-proteasome system for selective protein degradation
  • Molecular weight: 427.60
  • Molecular formula: C₂₁H₃₇N₃O₄S
  • Purity: ≥97.0%
  • Appearance: Solid, white to off-white
  • Storage at 4°C under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-40662


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.