Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) AffiniPure Goat Anti-Mouse IgG, Fcγ fragment specific, 1.5ml
SDP
Supplier:  Jackson Immuno Research Labs 115065071
Encompass_Preferred
Biotin-SP-AffiniPure Goat Anti-Mouse IgG, Fc Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9595436