Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) IgG Fraction Monoclonal Mouse Anti-Fluorescein (FITC), 0.5 mL
SDP
Supplier:  Jackson Immuno Research Labs 200062037
Encompass_Preferred
IgG fraction Monoclonal Mouse Anti-Fluorescein may be used either as direct conjugates, or for more sensitivity, they can be used unconjugated followed by a conjugated anti-mouse IgG (H+L) for signal enhancement.
Catalog No. NC1609997