Learn More
Medchemexpress LLC Biotin (Standard) | 58-85-5 | 99.8% | 250 MG
Supplier: Medchemexpress LLC 250MGHYB0511R
Biotin (Standard) is the analytical standard of Biotin. Biotin, also known as vitamin B7, serves as a coenzyme for five carboxylases in humans and is involved in the synthesis of fatty acids, isoleucine, and valine, as well as in gluconeogenesis. It is essential for cell growth, fatty acid production, and the metabolism of fats and amino acids.
- Analytical standard of Biotin
- Serves as a coenzyme for five carboxylases in humans
- Involved in the synthesis of fatty acids, isoleucine, and valine
- Involved in gluconeogenesis
- Essential for cell growth, fatty acid production, and metabolism of fats and amino acids
- Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS