Learn More
Medchemexpress LLC Biotin-TEG-ATFBA | 1264662-85-2 | C27H37F4N7O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516725MG
Encompass_Preferred
Biotin-TEG-ATFBA is a click chemistry reagent featuring a perfluorophenylazide group. It forms a stable azene intermediate that undergoes insertion and addition reactions with good yields after photolysis. The compound also contains an azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) and ring strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Contains a perfluorophenylazide group
  • Forms a highly stable azene intermediate
  • Undergoes insertion and addition reactions after photolysis
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Engages in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-73438