Learn More
Medchemexpress LLC Biotin-TEG-ATFBA | 1264662-85-2 | MFCD13184978 | 99.1% | 663.68 | C27H37F4N7O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15167210MG
Encompass_Preferred
Biotin-TEG-ATFBA is a biotin-labeled click chemistry reagent containing a perfluorophenylazide group. Upon photolysis it forms a stable azene intermediate that participates in insertion and cycloaddition reactions, enabling bioconjugation and linker construction for research applications. The compound is supplied as a solid with high purity and is intended for laboratory use.
Catalog No. 50-002-86497