Learn More
Medchemexpress LLC Biotin-VAD-FMK | 1135688-15-1 | ≥98.0% | 672.81 | C30H49FN6O8S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10089425MG
Encompass_Preferred
Biotin-VAD-FMK is a cell-permeable, irreversible biotin-labeled caspase inhibitor used to detect and label active caspases in cell lysates. The biotin tag enables affinity capture for pull-down assays and biochemical profiling of active caspases.
Catalog No. 50-002-11161