Learn More
Medchemexpress LLC Biotin-Vitamin B12 (Biotin-Cyanocobalamin B12) | 98.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0315A5MG
Encompass_Preferred
Biotin-Vitamin B12 (Biotin-Cyanocobalamin B12) is a biotinylated form of Vitamin B12, crucial for brain and nervous system function, and blood formation. It protects against oxidative stress and aids in inflammatory diseases. Involved in cellular metabolism, DNA synthesis, fatty acid synthesis, and energy production, it is valuable for research into vitamin B12 transport, cellular uptake, targeted delivery, and analytical detection.
  • Biotinylated vitamin B12
  • Supports brain and nervous system function
  • Essential for blood formation
  • Protects against oxidative-stress-associated pathologies
  • Beneficial in inflammatory diseases
  • Involved in cellular metabolism, DNA synthesis, and energy production
  • Facilitates research on vitamin B12 transport and cellular uptake
  • Supports targeted delivery and analytical detection research
Catalog No. 50-004-05825