Learn More
Medchemexpress LLC Biotin-X-NTA | 856661-92-2 | 98.1% | 715.98 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1471695MG
Encompass_Preferred
Biotin-X-NTA is a reagent used for detecting histidine-tagged proteins immobilized on nitrocellulose membranes.
  • Fluorescent dye
  • Labeling chemistry reagent
  • Functional group type biotin dye
  • Detects histidine-tagged proteins
  • Suitable for nitrocellulose membranes
Catalog No. 50-003-63617