Learn More
Biotium Biotin-XX, Succinimidyl Ester (6-((6-((B
Supplier:  Biotium 90052
Encompass_Preferred
Biotin-XX succinimidyl ester (6-((6-((Biotinoyl)amino)hexanoyl)amino)hexanoic acid succinimidyl ester) is readily coupled to a primary or secondary amine under mild conditions. The longer XX arm facilitates the interaction between biotin and avidin or streptavidin. Properties: White solid soluble in DMSO and moderately soluble in DMF; Store at -20°C; C26H41N5O7S; MW: 567.7; [89889-52-1]
Catalog No. 50-196-4809