Learn More
CPC Scientific Biotinyl-Phe-Asn-Ala-Pro-Phe-Asp-Val-Gly-Ile-Lys-Leu-Ser-Gly-Ala-Gln-Tyr-Gln-Gln-His-Gly-Arg-Ala-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OBST004B
Encompass
Suppressor of Food Intake and Gastric EmptyingONE-LETTER SEQUENCE: Biotinyl-FNAPFDVGIKLSGAQYQQHGRAL-NH2MOLECULAR FORMULA: C124H188N36O33S1MOLECULAR WEIGHT:2743.15STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-11-1], RESEARCH AREA: GastrointestinalREFERENCES: J.V.Zhang et al., Science, 310, 996 (2005); M. Kojima, H. Hosoda, Y. Date, M. Nakazato, H. Matsuo, and K. Kangawa, Nature, 402, 656 (1999)
Catalog No. 50-210-9971