Learn More
AdipoGen Biotinyl tyramide, 41994-02-9, MFCD27953026, 100 mg
SDP
Supplier:  AdipoGen CDXB0270M100
Encompass_Preferred
MF: C18H25N3O3S, MW: 363.5, ≥97%, Appearance: White to light pink solid. Biotin derivative. Substrate of the horseradish peroxidase enzyme and used as a reagent to amplify immunohistochemical signals. It is based on the HRP-catalyzed deposition of tyramide conjugates (such as biotinyl-tyramide) on a solid phase.
Catalog No. 50-101-6508