Learn More
Selleck Chemical LLC Biotinyl Tyramide
SDP
Supplier:  Selleck Chemical LLC S63185MG
Encompass_Preferred
Biotinyl Tyramide is a biotin derivative used for signal amplification in IHC and fluorescent in situ hybridization (ISH)
Catalog No. 50-000-03340