Learn More
American Research Products Inc Biotinylated AntiCSF1R
Supplier:  American Research Products Inc HDBS0265
Encompass
Biotinylated Anti-CSF1R (Cabiralizumab biosimilar) mAb is a biosimilar antibody with human reactivity produced in humanized Available in 100 g size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized antibodies are shipped at ambient temperature
Catalog No. 50-003-05108